Humic acid, sodium salt, 45-70%, Thermo Scientific Chemicals
Humic acid, sodium salt, 45-70%, Thermo Scientific Chemicals
Humic acid, sodium salt, 45-70%, Thermo Scientific Chemicals
Humic acid, sodium salt, 45-70%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Humic acid, sodium salt, 45-70%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
1 kg
Packaging:
View all
Glass bottle
Catalog number ACR120860050
Price (MYR)
375.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
375.00
Add to cart
Specifications
Chemical Name or MaterialHumic acid, sodium salt
Name Notetech., 50-60% (as humic acid)
CAS68131-04-4
Melting Point>300.0°C
ColorBlack