Humic acid, sodium salt, 45-70%, Thermo Scientific Chemicals
Humic acid, sodium salt, 45-70%, Thermo Scientific Chemicals
Humic acid, sodium salt, 45-70%, Thermo Scientific Chemicals
Humic acid, sodium salt, 45-70%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Humic acid, sodium salt, 45-70%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 g
100 g
Packaging:
View all
Plastic bottle
Catalog number ACR120860010
Price (MYR)
1,462.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,462.00
Add to cart
Chemical Identifiers
CAS7773-01-5
IUPAC Namemanganese(2+) dichloride
Molecular FormulaCl2Mn
InChI KeyGLFNIEUTAYBVOC-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Mn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Flakes or beads
Water=<1 %
Appearance (Color)Pink
Nickel (Ni)=<200 ppm
Iron (Fe)=<200 ppm