1,6-Hexanediol, 97%, Thermo Scientific Chemicals
1,6-Hexanediol, 97%, Thermo Scientific Chemicals
1,6-Hexanediol, 97%, Thermo Scientific Chemicals
1,6-Hexanediol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,6-Hexanediol, 97%, Thermo Scientific Chemicals
CAS: 629-11-8 | C6H14O2 | 118.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
1 kg
Packaging:
Glass bottle
Catalog number ACR120650025
Price (MYR)
1,096.00
Add to cart
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,096.00
Add to cart
Specifications
Chemical Name or Material1, 6-Hexanediol
Acidity0.1 mg KOH/g max.
CAS629-11-8
Melting Point40.0°C to 43.0°C