Hexamethylenetetramine, 99%, Thermo Scientific Chemicals
Hexamethylenetetramine, 99%, Thermo Scientific Chemicals
Hexamethylenetetramine, 99%, Thermo Scientific Chemicals
Hexamethylenetetramine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hexamethylenetetramine, 99%, Thermo Scientific Chemicals
CAS: 100-97-0 | C6H12N4 | 140.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
1 kg
Packaging:
Plastic Bottle
Catalog number ACR120610025
Price (MYR)
764.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
764.00
Add to cart
Chemical Identifiers
CAS768-66-1
IUPAC Name2,2,6,6-tetramethylpiperidine
Molecular FormulaC9H19N
InChI KeyRKMGAJGJIURJSJ-UHFFFAOYSA-N
SMILESCC1(C)CCCC(C)(C)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow to light brown
Refractive index1.4425 to 1.4465 (20°C, 589 nm)