Hexamethylenetetramine, 99%, Thermo Scientific Chemicals
Hexamethylenetetramine, 99%, Thermo Scientific Chemicals
Hexamethylenetetramine, 99%, Thermo Scientific Chemicals
Hexamethylenetetramine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hexamethylenetetramine, 99%, Thermo Scientific Chemicals
CAS: 100-97-0 | C6H12N4 | 140.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
Packaging:
Plastic Bottle
Catalog number ACR120610010
Price (MYR)
599.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
599.00
Add to cart
Chemical Identifiers
CAS13463-67-7
IUPAC Namedioxotitanium
Molecular FormulaO2Ti
InChI KeyGWEVSGVZZGPLCZ-UHFFFAOYSA-N
SMILESO=[Ti]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Elemental AnalysisTitanium Ti: ≥58.95% (Theory 59.934%)
Total Metal Impurities≤0.0055% (≤55ppm)