Hexamethyleneimine, 99%, Thermo Scientific Chemicals
Hexamethyleneimine, 99%, Thermo Scientific Chemicals
Hexamethyleneimine, 99%, Thermo Scientific Chemicals
Hexamethyleneimine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hexamethyleneimine, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
Packaging:
Glass bottle
Catalog number ACR120590250
Price (MYR)
365.00
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
365.00
Add to cart
Specifications
Chemical Name or MaterialHexamethyleneimine
CAS111-49-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Health Hazard 3GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre