n-Heptane, 99+%, pure, Thermo Scientific Chemicals
n-Heptane, 99+%, pure, Thermo Scientific Chemicals
n-Heptane, 99+%, pure, Thermo Scientific Chemicals
n-Heptane, 99+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Heptane, 99+%, pure, Thermo Scientific Chemicals
CAS: 142-82-5 | C7H16 | 100.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
Catalog number ACR120340050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 L
Request bulk or custom format
Specifications
Chemical Name or Materialn-Heptane
Melting Point-91.0°C
CAS142-82-5
CAS Min %99.0