Guanidine hydrochloride, 98%, with <0.4% anticaking agent (SiO2), Thermo Scientific Chemicals
Guanidine hydrochloride, 98%, with &lt;0.4% anticaking agent (SiO<sub>2</sub>), Thermo Scientific Chemicals
Guanidine hydrochloride, 98%, with &lt;0.4% anticaking agent (SiO<sub>2</sub>), Thermo Scientific Chemicals
Guanidine hydrochloride, 98%, with &lt;0.4% anticaking agent (SiO<sub>2</sub>), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Guanidine hydrochloride, 98%, with <0.4% anticaking agent (SiO2), Thermo Scientific Chemicals
Guanidine hydrochloride, 98%, with <0.4% anticaking agent (SiO2), ACROS Organics, Quantity: 1kg, Packaging: Plastic bottle, White to Yellow, Melting Point: 179.0 deg.C to 186.0 deg.C, Molecular Weight: 95.53, Percent Purity: 98%, Assay Percent Range: 0.5% max. Ammonium chloride (Argentometry)
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number ACR120232500
Price (MYR)
407.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
407.00
Add to cart
Specifications
Chemical Name or MaterialGuanidine hydrochloride
CAS50-01-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
Health Hazard 3GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse