Guanidine hydrochloride, 98%, with <0.4% anticaking agent (SiO2), Thermo Scientific Chemicals
Guanidine hydrochloride, 98%, with &lt;0.4% anticaking agent (SiO<sub>2</sub>), Thermo Scientific Chemicals
Guanidine hydrochloride, 98%, with &lt;0.4% anticaking agent (SiO<sub>2</sub>), Thermo Scientific Chemicals
Guanidine hydrochloride, 98%, with &lt;0.4% anticaking agent (SiO<sub>2</sub>), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Guanidine hydrochloride, 98%, with <0.4% anticaking agent (SiO2), Thermo Scientific Chemicals
Guanidine hydrochloride, 98%, with <0.4% anticaking agent (SiO2), ACROS Organics, Quantity: 1kg, Packaging: Plastic bottle, White to Yellow, Melting Point: 179.0 deg.C to 186.0 deg.C, Molecular Weight: 95.53, Percent Purity: 98%, Assay Percent Range: 0.5% max. Ammonium chloride (Argentometry)
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Plastic bottle
Catalog number ACR120230010
Price (MYR)
691.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
691.00
Add to cart
Chemical Identifiers
CAS37380-43-1
MDL NumberMFCD00132705
SpecificationsSpecification SheetSpecification Sheet
TOC-AFNOR test=<10 ppm (total)
Appearance (Color)White to pink-beige
Moisture holding capacity61.0 to 69.0 %
Harmonic mean size0.430 to 0.690 mm
Particle size=<8.0 % (> 1.180 mm)