Glycidol, 96%, Thermo Scientific Chemicals
Glycidol, 96%, Thermo Scientific Chemicals
Glycidol, 96%, Thermo Scientific Chemicals
Glycidol, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glycidol, 96%, Thermo Scientific Chemicals
CAS: 556-52-5 | C3H6O2 | 74.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number ACR120051000
Price (MYR)
648.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
648.00
Add to cart
Chemical Identifiers
CAS20989-17-7
IUPAC Name(2S)-2-amino-2-phenylethan-1-ol
Molecular FormulaC8H11NO
InChI KeyIJXJGQCXFSSHNL-MRVPVSSYSA-N
SMILESN[C@H](CO)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystalline powder
Melting Point (clear melt)74-80°C
Solution TestClear (2.5% in 1N HCl)
Appearance (Color)White to pale yellow
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0%