2-Furoic acid, 98%, Thermo Scientific Chemicals
2-Furoic acid, 98%, Thermo Scientific Chemicals
2-Furoic acid, 98%, Thermo Scientific Chemicals
2-Furoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Furoic acid, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number ACR119821000
Price (MYR)
522.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
522.00
Add to cart
Chemical Identifiers
CAS32916-51-1
IUPAC Namechloro(cyclopentyl)magnesium
Molecular FormulaC5H9ClMg
InChI KeyGROGEABZCKQTJW-UHFFFAOYSA-M
SMILESCl[Mg]C1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Grignard1.9 to 2.2 M
Appearance (Color)Clear to hazy yellow to grey to brown
AppearanceMay develop some turbidity or precipitate
Appearance (Form)Solution