Furfurylamine, 99+%, Thermo Scientific Chemicals
Furfurylamine, 99+%, Thermo Scientific Chemicals
Furfurylamine, 99+%, Thermo Scientific Chemicals
Furfurylamine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Furfurylamine, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
250 mL
Packaging:
Glass bottle
Catalog number ACR119800050
Price (MYR)
388.00
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
388.00
Add to cart
Specifications
Chemical Name or MaterialFurfurylamine
Name Note99+%
CAS617-89-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Flammable liquid and vapor.