Formamidine acetate, 99%, Thermo Scientific Chemicals
Formamidine acetate, 99%, Thermo Scientific Chemicals
Formamidine acetate, 99%, Thermo Scientific Chemicals
Formamidine acetate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Formamidine acetate, 99%, Thermo Scientific Chemicals
CAS: 3473-63-0 | CH4N2·C2H4O2 | 104.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number ACR119711000
Price (MYR)
529.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
529.00
Add to cart
Specifications
Chemical Name or MaterialFormamidine acetate
Name Note99%
CAS3473-63-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: