1-Fluoro-2-nitrobenzene, 99%, Thermo Scientific Chemicals
1-Fluoro-2-nitrobenzene, 99%, Thermo Scientific Chemicals
1-Fluoro-2-nitrobenzene, 99%, Thermo Scientific Chemicals
1-Fluoro-2-nitrobenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Fluoro-2-nitrobenzene, 99%, Thermo Scientific Chemicals
CAS: 1493-27-2 | C6H4FNO2 | 141.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
Packaging:
Glass bottle
Catalog number ACR119480100
Price (MYR)
378.00
Add to cart
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
378.00
Add to cart
Chemical Identifiers
CAS7789-23-3
IUPAC Namepotassium fluoride
Molecular FormulaFK
InChI KeyNROKBHXJSPEDAR-UHFFFAOYSA-M
SMILES[F-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration99.0 to 101.0 %
Free acid (HF)=<0.1 %
Potassium fluosilicate=<0.1 %
Sulfate (SO4)=<200 ppm
Cadmium (Cd)=<5 ppm