Flavone, 99+%, Thermo Scientific Chemicals
Flavone, 99+%, Thermo Scientific Chemicals
Flavone, 99+%, Thermo Scientific Chemicals
Flavone, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Flavone, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number ACR119160250
Price (MYR)
2,307.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,307.00
Add to cart
Chemical Identifiers
CAS1327-53-3
IUPAC Namediarsorosooxidane
Molecular FormulaAs2O3
InChI KeyIKWTVSLWAPBBKU-UHFFFAOYSA-N
SMILESO=[As]O[As]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.