Flavone, 99+%, Thermo Scientific Chemicals
Flavone, 99+%, Thermo Scientific Chemicals
Flavone, 99+%, Thermo Scientific Chemicals
Flavone, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Flavone, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR119160050
Price (MYR)
723.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
723.00
Add to cart
Chemical Identifiers
CAS140-75-0
IUPAC Name1-(4-fluorophenyl)methanamine
Molecular FormulaC7H8FN
InChI KeyIIFVWLUQBAIPMJ-UHFFFAOYSA-N
SMILESNCC1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5115 to 1.5155 (20°C, 589 nm)
GC>=96.0 %