Farnesol, 96%, mixture of isomers, Thermo Scientific Chemicals
Farnesol, 96%, mixture of isomers, Thermo Scientific Chemicals
Farnesol, 96%, mixture of isomers, Thermo Scientific Chemicals
Farnesol, 96%, mixture of isomers, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Farnesol, 96%, mixture of isomers, Thermo Scientific Chemicals
CAS: 4602-84-0 | C15H26O | 222.37 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR11912025025 mLGlass bottle
Catalog number ACR119120250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS579-74-8
IUPAC Name1-(2-methoxyphenyl)ethan-1-one
Molecular FormulaC9H10O2
InChI KeyDWPLEOPKBWNPQV-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear light yellow to orange to light brown
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Liquid
Refractive index1.5383 to 1.5403 (20°C, 589 nm)