Ethylene glycol dimethyl ether, 99+%, extra pure, stabilized with BHT, Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99+%, extra pure, stabilized with BHT, Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99+%, extra pure, stabilized with BHT, Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99+%, extra pure, stabilized with BHT, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99+%, extra pure, stabilized with BHT, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
Packaging:
Glass Bottle
Catalog number ACR118450025
Price (MYR)
1,397.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,397.00
Add to cart
Chemical Identifiers
CAS112-18-5
IUPAC Namedodecyldimethylamine
Molecular FormulaC14H31N
InChI KeyYWFWDNVOPHGWMX-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Titration>=94 % (Tertiary amine)
GC>=94 % (Dodecyl)
Appearance (Color)Clear to slightly hazy