Esculin hydrate, 97%, Thermo Scientific Chemicals
Esculin hydrate, 97%, Thermo Scientific Chemicals
Esculin hydrate, 97%, Thermo Scientific Chemicals
Esculin hydrate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Esculin hydrate, 97%, Thermo Scientific Chemicals
CAS: 531-75-9 | C15H16O9 | 340.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
5 g
50 g
Packaging:
Glass bottle
Catalog number ACR117830100
Price (MYR)
577.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
577.00
Add to cart
Specifications
Chemical Name or MaterialEsculin hydrate
CAS531-75-9
Melting Point203.0°C to 205.0°C
EINECS Number208-517-5
Infrared SpectrumAuthentic