Epichlorohydrin, 99%, Thermo Scientific Chemicals
Epichlorohydrin, 99%, Thermo Scientific Chemicals
Epichlorohydrin, 99%, Thermo Scientific Chemicals
Epichlorohydrin, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Epichlorohydrin, 99%, Thermo Scientific Chemicals
CAS: 106-89-8 | C3H5ClO | 92.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number ACR117780250
Price (MYR)
703.00
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
703.00
Add to cart
Chemical Identifiers
CAS944721-52-2
IUPAC Name2,3,4,5,6-pentafluorophenyl 11-(trimethoxysilyl)undecanoate
Molecular FormulaC20H29F5O5Si
InChI KeyKSYPNQIBGIOYPZ-UHFFFAOYSA-N
SMILESCO[Si](CCCCCCCCCCC(=O)OC1=C(F)C(F)=C(F)C(F)=C1F)(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥94.0%
Refractive Index1.4360-1.4420 @ 20?C