Epichlorohydrin, 99%, Thermo Scientific Chemicals
Epichlorohydrin, 99%, Thermo Scientific Chemicals
Epichlorohydrin, 99%, Thermo Scientific Chemicals
Epichlorohydrin, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Epichlorohydrin, 99%, Thermo Scientific Chemicals
CAS: 106-89-8 | C3H5ClO | 92.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
25 mL
1 L
Packaging:
Glass bottle
Catalog number ACR117780025
Price (MYR)
2,526.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,526.00
Add to cart
Specifications
Chemical Name or MaterialEpichlorohydrin
CAS106-89-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause cancer.
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
May cause an allergic skin reaction.
Flammable liquid and vapor.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with