Epichlorohydrin, 99%, Thermo Scientific Chemicals
Epichlorohydrin, 99%, Thermo Scientific Chemicals
Epichlorohydrin, 99%, Thermo Scientific Chemicals
Epichlorohydrin, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Epichlorohydrin, 99%, Thermo Scientific Chemicals
CAS: 106-89-8 | C3H5ClO | 92.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
25 mL
2.5 L
Packaging:
Glass bottle
Catalog number ACR117780010
Price (MYR)
951.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
951.00
Add to cart
Chemical Identifiers
CAS269410-22-2
IUPAC Name2-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Molecular FormulaC13H19BO4
InChI KeyWFSJROCEOJANPD-UHFFFAOYSA-N
SMILESCOC1=CC(=CC=C1O)B1OC(C)(C)C(C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=96.0 %
Melting point101°C to 106°C