Ellagic acid, 97%
Ellagic acid, 97%
Ellagic acid, 97%
Thermo Scientific Chemicals
Ellagic acid, 97%
CAS: 476-66-4 | C14H6O8 | 302.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
2.5 g
10 g
Packaging:
Glass bottle
Catalog number ACR117740500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialEllagic acid
CAS476-66-4
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.