Ellagic acid, 97%
Ellagic acid, 97%
Ellagic acid, 97%
Thermo Scientific Chemicals
Ellagic acid, 97%
CAS: 476-66-4 | C14H6O8 | 302.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass bottle
Catalog number ACR117740025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS142-03-0
IUPAC Name(acetyloxy)(hydroxy)alumanyl acetate
Molecular FormulaC4H7AlO5
InChI KeyHQQUTGFAWJNQIP-UHFFFAOYSA-K
SMILESCC(=O)O[Al](O)OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay from Suppliers CofA≥98.0% (U.S. specification)
CommentAl2O3 content ≥ 28.0% (non-U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated