Dithiooxamide, 98%, Thermo Scientific Chemicals
Dithiooxamide, 98%, Thermo Scientific Chemicals
Dithiooxamide, 98%, Thermo Scientific Chemicals
Dithiooxamide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dithiooxamide, 98%, Thermo Scientific Chemicals
CAS: 79-40-3 | C2H4N2S2 | 120.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number ACR117550250
Price (MYR)
1,670.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,670.00
Add to cart
Chemical Identifiers
CAS51012-65-8
IUPAC Name2-bromo-1-(2-methylphenyl)ethan-1-one
Molecular FormulaC9H9BrO
InChI KeyXMGAXELQRATLJP-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1C(=O)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%