Dithiooxamide, 98%, Thermo Scientific Chemicals
Dithiooxamide, 98%, Thermo Scientific Chemicals
Dithiooxamide, 98%, Thermo Scientific Chemicals
Dithiooxamide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dithiooxamide, 98%, Thermo Scientific Chemicals
CAS: 79-40-3 | C2H4N2S2 | 120.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number ACR117550050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size0.02 Microns
Concentration (weight/weight)SiO₂ 40% min