5,5'-Dithiobis-(2-nitrobenzoic acid), 99%, Thermo Scientific Chemicals
5,5'-Dithiobis-(2-nitrobenzoic acid), 99%, Thermo Scientific Chemicals
5,5'-Dithiobis-(2-nitrobenzoic acid), 99%, Thermo Scientific Chemicals
5,5'-Dithiobis-(2-nitrobenzoic acid), 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5,5'-Dithiobis-(2-nitrobenzoic acid), 99%, Thermo Scientific Chemicals
CAS: 69-78-3 | C14H8N2O8S2 | 396.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
25 g
Packaging:
View all
Catalog number ACR117540050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS6410-10-2
IUPAC Name(1Z)-1-[2-(4-nitrophenyl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one
Molecular FormulaC16H11N3O3
InChI KeyACHQDMGGRZPFAW-VLGSPTGOSA-N
SMILES[O-][N+](=O)C1=CC=C(NN=C2/C(=O)C=CC3=CC=CC=C23)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
lambda-max (UV-Vis)485-491 nm (in Toluene)
Solution Test0.1% (w/v) in Acetone : Clear
Appearance (Color)Red