2,5-Diphenyloxazole, Scintillation Grade 99%, Thermo Scientific Chemicals
2,5-Diphenyloxazole, Scintillation Grade 99%, Thermo Scientific Chemicals
2,5-Diphenyloxazole, Scintillation Grade 99%, Thermo Scientific Chemicals
2,5-Diphenyloxazole, Scintillation Grade 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,5-Diphenyloxazole, Scintillation Grade 99%, Thermo Scientific Chemicals
CAS: 92-71-7 | C15H11NO | 221.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic Bottle
Catalog number ACR117381000
Price (MYR)
962.00
Add to cart
Quantity:
100 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
962.00
Add to cart
Specifications
Chemical Name or Material2, 5-Diphenyloxazole
Melting Point70°C to 72°C
Name Note99%
CAS92-71-7
Physical FormCrystalline Powder
View more
for fluorescence/luminescence spectroscopy
RUO – Research Use Only