9,10-Diphenylanthracene, 98%, Thermo Scientific Chemicals
9,10-Diphenylanthracene, 98%, Thermo Scientific Chemicals
9,10-Diphenylanthracene, 98%, Thermo Scientific Chemicals
9,10-Diphenylanthracene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
9,10-Diphenylanthracene, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number ACR117200050
Price (MYR)
1,346.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,346.00
Add to cart
Chemical Identifiers
CAS14937-45-2
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline or fused solid, lumps or powder
Identification (FTIR)Conforms
Melting Point (clear melt)56-63°C
Appearance (Color)White to cream or pale yellow
Assay (Titration ex Bromide)≥98.0 to ≤102.0%