9,10-Diphenylanthracene, 98%, Thermo Scientific Chemicals
9,10-Diphenylanthracene, 98%, Thermo Scientific Chemicals
9,10-Diphenylanthracene, 98%, Thermo Scientific Chemicals
9,10-Diphenylanthracene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
9,10-Diphenylanthracene, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR117200010
Price (MYR)
540.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
540.00
Add to cart
Chemical Identifiers
CAS3125-64-2
IUPAC Name1-isothiocyanato-3-methoxybenzene
Molecular FormulaC8H7NOS
InChI KeyWHBYCPUKGYEYFU-UHFFFAOYSA-N
SMILESCOC1=CC=CC(=C1)N=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.6370-1.6410 @ 20?C
Assay (GC)>97.5%
Appearance (Color)Clear colorless to yellow