1,4-Dioxane, 99+%, extra pure, stabilized, Thermo Scientific Chemicals
1,4-Dioxane, 99+%, extra pure, stabilized, Thermo Scientific Chemicals
1,4-Dioxane, 99+%, extra pure, stabilized, Thermo Scientific Chemicals
1,4-Dioxane, 99+%, extra pure, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Dioxane, 99+%, extra pure, stabilized, Thermo Scientific Chemicals
CAS: 123-91-1 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
25 L
Packaging:
View all
Glass Bottle
Catalog number ACR117110010
Price (MYR)
661.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
661.00
Add to cart
Chemical Identifiers
CAS75-52-5
IUPAC Namenitromethane
Molecular FormulaCH3NO2
InChI KeyLYGJENNIWJXYER-UHFFFAOYSA-N
SMILESC[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<10 APHA
Infrared spectrumConforms
GC>=95 %
Water=<0.05 % (Coulometric)