Di-n-octylamine, 95%, Thermo Scientific Chemicals
Di-n-octylamine, 95%, Thermo Scientific Chemicals
Di-n-octylamine, 95%, Thermo Scientific Chemicals
Di-n-octylamine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Di-n-octylamine, 95%, Thermo Scientific Chemicals
CAS: 1120-48-5 | C16H35N | 241.46 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR117081000100 gGlass bottle
Catalog number ACR117081000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1600-27-7
IUPAC Namemercury(2+) diacetate
Molecular FormulaC4H6HgO4
InChI KeyBRMYZIKAHFEUFJ-UHFFFAOYSA-L
SMILES[Hg++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration Complexometric>=97.5 %
Sulfate (SO4)=<100 ppm
Appearance (Color)White to almost white
Melting point179°C to 182°C