3,5-Dimethylpyrazole, 99%, Thermo Scientific Chemicals
3,5-Dimethylpyrazole, 99%, Thermo Scientific Chemicals
3,5-Dimethylpyrazole, 99%, Thermo Scientific Chemicals
3,5-Dimethylpyrazole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,5-Dimethylpyrazole, 99%, Thermo Scientific Chemicals
CAS: 67-51-6 | C5H8N2 | 96.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number ACR116701000
Price (MYR)
617.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
617.00
Add to cart
Chemical Identifiers
CAS3761-92-0
IUPAC Namebromo(hexyl)magnesium
Molecular FormulaC6H13BrMg
InChI KeySFXUWNMDTWQSHE-UHFFFAOYSA-M
SMILESCCCCCC[Mg]Br
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Grignard0.6 to 1 M
Appearance (Color)Brown
Appearance (Form)Solution