2,6-Dimethylphenol, 99%, Thermo Scientific Chemicals
2,6-Dimethylphenol, 99%, Thermo Scientific Chemicals
2,6-Dimethylphenol, 99%, Thermo Scientific Chemicals
2,6-Dimethylphenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,6-Dimethylphenol, 99%, Thermo Scientific Chemicals
CAS: 576-26-1 | C8H10O | 122.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Glass bottle
Catalog number ACR116572500
Price (MYR)
452.00
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
452.00
Add to cart
Chemical Identifiers
CAS25016-01-7
IUPAC Name5-bromo-2-methoxybenzaldehyde
Molecular FormulaC8H7BrO2
InChI KeyIJIBRSFAXRFPPN-UHFFFAOYSA-N
SMILESCOC1=CC=C(Br)C=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to cream
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
Free acid (titration)≤1.0%
Melting Point (clear melt)112.5-118.5?C