N,O-Dimethylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
N,O-Dimethylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
N,O-Dimethylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
N,O-Dimethylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,O-Dimethylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
CAS: 6638-79-5 | C2H8ClNO | 97.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number ACR116340250
Price (MYR)
776.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
776.00
Add to cart
Chemical Identifiers
CAS3094-87-9
IUPAC Nameλ²-iron(2+) diacetate
Molecular FormulaC4H6FeO4
InChI KeyLNOZJRCUHSPCDZ-UHFFFAOYSA-L
SMILES[Fe++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration with KMnO4>=94 %
Appearance (Color)Off-white to light grey to light brown
Infrared spectrumConforms
Coupling reagent
RUO – Research Use Only