N,O-Dimethylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
N,O-Dimethylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
N,O-Dimethylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
N,O-Dimethylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,O-Dimethylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
CAS: 6638-79-5 | C2H8ClNO | 97.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR116340050
Price (MYR)
444.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
444.00
Add to cart
Chemical Identifiers
CAS10199-89-0
IUPAC Name4-chloro-7-nitro-2,1,3-benzoxadiazole
Molecular FormulaC6H2ClN3O3
InChI KeyIGHBXJSNZCFXNK-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(Cl)C2=NON=C12
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)Yellow to pale brown
Assay (GC)≥97.5%
Melting Point (clear melt)96-101?C
Coupling reagent
RUO – Research Use Only