5,5-Dimethylhydantoin, 97%, Thermo Scientific Chemicals
5,5-Dimethylhydantoin, 97%, Thermo Scientific Chemicals
5,5-Dimethylhydantoin, 97%, Thermo Scientific Chemicals
5,5-Dimethylhydantoin, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5,5-Dimethylhydantoin, 97%, Thermo Scientific Chemicals
CAS: 77-71-4 | C5H8N2O2 | 128.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Plastic bottle
Catalog number ACR116312500
Price (MYR)
533.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
533.00
Add to cart
Chemical Identifiers
CAS10534-89-1
IUPAC Namecobalt(3+) hexaamine trichloride
Molecular FormulaCl3CoH18N6
InChI KeyJXBGZYGSWFSYFI-UHFFFAOYSA-K
SMILESN.N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Co+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)96.1% min