4-Dimethylaminocinnamaldehyde, 98%
4-Dimethylaminocinnamaldehyde, 98%
4-Dimethylaminocinnamaldehyde, 98%
4-Dimethylaminocinnamaldehyde, 98%
Thermo Scientific Chemicals
4-Dimethylaminocinnamaldehyde, 98%
CAS: 6203-18-5 | C11H13NO | 175.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number ACR115790250
Price (MYR)
1,671.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,671.00
Add to cart
Chemical Identifiers
CAS142-73-4
IUPAC Name2-[(carboxymethyl)amino]acetic acid
Molecular FormulaC4H7NO4
InChI KeyNBZBKCUXIYYUSX-UHFFFAOYSA-N
SMILESOC(=O)CNCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)White or cream to pale brown
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%