4-Dimethylaminocinnamaldehyde, 98%
4-Dimethylaminocinnamaldehyde, 98%
4-Dimethylaminocinnamaldehyde, 98%
4-Dimethylaminocinnamaldehyde, 98%
Thermo Scientific Chemicals
4-Dimethylaminocinnamaldehyde, 98%
CAS: 6203-18-5 | C11H13NO | 175.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR115790050
Price (MYR)
666.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
666.00
Add to cart
Chemical Identifiers
CAS7789-20-0
IUPAC Name[(²H)oxy](²H)
Molecular FormulaH2O
InChI KeyXLYOFNOQVPJJNP-ZSJDYOACSA-N
SMILES[2H]O[2H]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
D-Enrichment>=99.75 %
Appearance (Color)Clear colorless