4-Dimethylaminobenzoic acid, 98%, Thermo Scientific Chemicals
4-Dimethylaminobenzoic acid, 98%, Thermo Scientific Chemicals
4-Dimethylaminobenzoic acid, 98%, Thermo Scientific Chemicals
4-Dimethylaminobenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Dimethylaminobenzoic acid, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number ACR115760250
Price (MYR)
486.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
486.00
Add to cart
Specifications
Chemical Name or Material4-Dimethylaminobenzoic acid
CAS619-84-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0