N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number ACR115690050
Price (MYR)
453.00
Add to cart
Quantity:
5 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
453.00
Add to cart
Chemical Identifiers
CAS109-52-4
IUPAC Namepentanoic acid
Molecular FormulaC5H10O2
InChI KeyNQPDZGIKBAWPEJ-UHFFFAOYSA-N
SMILESCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Water=<0.1 %