N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
5 mL
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number ACR115690010
Price (MYR)
563.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
563.00
Add to cart
Specifications
Chemical Name or MaterialN, N-Dimethylacetamide
CAS127-19-5
CAS Min %98.5
Health Hazard 1Signal Word: WARNING!
CAS Max %100.0