3,5-Dimethoxy-4-hydroxycinnamic acid, 98%, predominantly trans isomer, Thermo Scientific Chemicals
CAS: 530-59-6 | C11H12O5 | 224.21 g/mol
Have Questions?
Change view
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number ACR115550050
Price (MYR)
722.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
722.00
Add to cart
Specifications
Chemical Name or Material3, 5-Dimethoxy-4-hydroxycinnamic acid
CAS530-59-6
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0