3,4-Dimethoxybenzoic acid, 99+%, Thermo Scientific Chemicals
3,4-Dimethoxybenzoic acid, 99+%, Thermo Scientific Chemicals
3,4-Dimethoxybenzoic acid, 99+%, Thermo Scientific Chemicals
3,4-Dimethoxybenzoic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,4-Dimethoxybenzoic acid, 99+%, Thermo Scientific Chemicals
CAS: 93-07-2 | C9H10O4 | 182.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number ACR115451000
Price (MYR)
598.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
598.00
Add to cart
Chemical Identifiers
CAS93-07-2
IUPAC Name3,4-dimethoxybenzoic acid
Molecular FormulaC9H10O4
InChI KeyDAUAQNGYDSHRET-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1OC)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point180°C to 184°C
HPLC>=99.0 %