3,4-Dimethoxybenzoic acid, 99+%, Thermo Scientific Chemicals
3,4-Dimethoxybenzoic acid, 99+%, Thermo Scientific Chemicals
3,4-Dimethoxybenzoic acid, 99+%, Thermo Scientific Chemicals
3,4-Dimethoxybenzoic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,4-Dimethoxybenzoic acid, 99+%, Thermo Scientific Chemicals
CAS: 93-07-2 | C9H10O4 | 182.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR115450250
Price (MYR)
403.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
403.00
Add to cart
Chemical Identifiers
CAS112-92-5
IUPAC Nameoctadecan-1-ol
Molecular FormulaC18H38O
InChI KeyGLDOVTGHNKAZLK-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder or flakes or pellets
Infrared spectrumConforms
GC>=94 %
Melting point55°C to 60°C