2,4-Dimethoxyaniline, 97%, Thermo Scientific™
2,4-Dimethoxyaniline, 97%, Thermo Scientific™
2,4-Dimethoxyaniline, 97%, Thermo Scientific™
2,4-Dimethoxyaniline, 97%, Thermo Scientific™
2,4-Dimethoxyaniline, 97%, Thermo Scientific™
Have Questions?
Catalog NumberQuantityPackaging
ACR11535050050 gGlass bottle
Catalog number ACR115350500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 4-Dimethoxyaniline
Name Note97%
CAS2735-04-8
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
View more
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
RUO – Research Use Only