2,5-Dimercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
2,5-Dimercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
2,5-Dimercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
2,5-Dimercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,5-Dimercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
View all
Plastic bottle
Catalog number ACR115311000
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS34592-47-7
IUPAC Name(4R)-1,3-thiazolidin-3-ium-4-carboxylate
Molecular FormulaC4H7NO2S
InChI KeyDZLNHFMRPBPULJ-VKHMYHEASA-N
SMILES[O-]C(=O)[C@@H]1CSC[NH2+]1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Optical Rotation-138° to -144° (c=1 in water)
FormCrystals or powder or crystalline powder or needles