2,5-Dimercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
2,5-Dimercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
2,5-Dimercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
2,5-Dimercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,5-Dimercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
View all
Plastic bottle
Catalog number ACR115310250
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS677-22-5
IUPAC Nametert-butyl(chloro)magnesium
Molecular FormulaC4H9ClMg
InChI KeyZDRJSYVHDMFHSC-UHFFFAOYSA-M
SMILESCC(C)(C)[Mg]Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark brown
Appearance (Form)Solution
Titration Active base24 to 26 %