2,5-Dimercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
2,5-Dimercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
2,5-Dimercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
2,5-Dimercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,5-Dimercapto-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
View all
Glass bottle
Catalog number ACR115310050
Price (MYR)
371.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
371.00
Add to cart
Specifications
Chemical Name or Material2,5-Dimercapto-1,3,4-thiadiazole
Name Note98%
CAS1072-71-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning