N,N-Diethyl-m-toluamide, 98%, Thermo Scientific Chemicals
N,N-Diethyl-m-toluamide, 98%, Thermo Scientific Chemicals
N,N-Diethyl-m-toluamide, 98%, Thermo Scientific Chemicals
N,N-Diethyl-m-toluamide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Diethyl-m-toluamide, 98%, Thermo Scientific Chemicals
CAS: 134-62-3 | C12H17NO | 191.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number ACR114571000
Price (MYR)
389.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
389.00
Add to cart
Chemical Identifiers
CAS583-71-1
IUPAC Name4-bromo-1,2-dimethylbenzene
Molecular FormulaC8H9Br
InChI KeyQOGHRLGTXVMRLM-UHFFFAOYSA-N
SMILESCC1=CC=C(Br)C=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.5535-1.5585 @ 20?C
Assay (GC)≥96.0%